L-Tyrosine benzyl ester 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
H-Tyr-OBzl, Benzyl tyrosinate, (S)-Benzyl 2-amino-3-(4-hydroxyphenyl)propanoate, (S)-2-Amino-3-(4-hydroxyphenyl)propionic acid benzyl ester
A212289-100G
A212289-10G
A212289-1G
A212289-25G
A212289-5G
103818-186EA
21
USD
103818-186
103848-884
76825-930
76825-932
77087-350
77087-352
77087-354
L-Tyrosine benzyl ester 95%
L-Tyrosine benzyl ester
Caution: for Research Use Only
Formula:
C₁₆H₁₇NO₃ MW: 271.316 g/mol Boiling Pt: 438.7 °C (760 mm Hg) Flash Pt: 219.1 °C Storage Temperature: Freezer |
MDL Number:
MFCD00191034 CAS Number: 42406-77-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...