3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%

Supplier: Ambeed
Warning

A208507-100G A208507-1MG A208507-1G A208507-25G A208507-5G
103817-876EA 17 USD
103817-876 103848-562 77030-424 77030-426 77086-880 77086-882 77086-884
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT) 98%
3-(4,5-Dimethylthiazol-2-yl)-2,5-diphenyltetrazolium bromide (MTT)

Caution: for Research Use Only

Formula: C₁₈H₁₆N₅BrS
Melting Pt: 195 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00011964
CAS Number: 298-93-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR