4-Bromoindole-3-carboxyaldehyde 98%

Supplier: Ambeed
Warning

A196612-10G A196612-250MG A196612-5G A196612-1G
103816-858EA 19 USD
103816-858 103847-472 76825-424 76825-426 77085-436 77302-522
4-Bromoindole-3-carboxyaldehyde 98%
4-Bromoindole-3-carboxyaldehyde

Caution: for Research Use Only

Formula: C₉H₆BrNO
Boiling Pt: 395.6 °C (760 mm Hg)
Melting Pt: 179…180 °C
Flash Pt: 193 °C
Storage Temperature: Refrigerator
MDL Number: MFCD05864695
CAS Number: 98600-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR