L-(+)-Methyl-tert-leucinate hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A190165-25G
A190165-5G
A190165-100G
A190165-10G
A190165-1G
A190165-500G
103846-868EA
6
USD
103846-868
77084-656
77084-658
77111-722
77111-724
77111-726
77111-728
L-(+)-Methyl-tert-leucinate hydrochloride 97%
L(+)-Methyl tert-leucinate hydrochloride
Caution: for Research Use Only
Formula:
C₇H₁₅NO₂·HCl Melting Pt: 183 °C Flash Pt: 78.9 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00040604 CAS Number: 63038-27-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...