2-(Trifluoromethyl)benzophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Phenyl(2-(trifluoromethyl)phenyl)methanone
A189592-1G
A189592-5G
A189592-10G
103816-206EA
15
USD
103816-206
103846-800
77084-572
77084-574
77871-953
2-(Trifluoromethyl)benzophenone 98%
2-(Trifluoromethyl)benzophenone
Caution: for Research Use Only
Formula:
C₁₄H₉F₃O MW: 250.22 Boiling Pt: 369 °C (760 mm Hg) Melting Pt: 62 Flash Pt: 177 °C Storage Temperature: Ambient |
MDL Number:
MFCD00000377 CAS Number: 727-99-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...