Indole-3-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1H-Indole-3-carboxylic acid

A188858-25G A188858-100G A188858-100MG A188858-10G A188858-500G
103846-730EA 6 USD
103846-730 77084-482 77274-368 77274-370 77274-372 77274-374
Indole-3-carboxylic acid 98%
Indole-3-carboxylic acid

Caution: for Research Use Only

Formula: C₉H₇NO₂
Boiling Pt: 419.6 °C (760 mm Hg)
Melting Pt: 232…240 °C
Flash Pt: 207.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00005624
CAS Number: 771-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR