5-Bromo-2-iodotoluene 98%

Supplier: Ambeed
Warning

A186165-100G A186165-10G A186165-500G A186165-25G
76769-740EA 55 USD
76769-740 76769-742 76769-744 77084-150
5-Bromo-2-iodotoluene 98%
5-Bromo-2-iodotoluene

Caution: for Research Use Only

Formula: C₇H₆BrI
MDL Number: MFCD00060664
CAS Number: 116632-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR