2,4-Dichloro-5-methylpyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4-Dichloro-5-picoline
A183924-100MG
A183924-1G
A183924-25G
A183924-5G
A183924-10G
103815-686EA
8
USD
103815-686
103846-248
76974-540
77083-858
77083-860
77083-862
77611-788
2,4-Dichloro-5-methylpyridine 98%
2,4-Dichloro-5-methylpyridine
Caution: for Research Use Only
Formula:
C₆H₅Cl₂N MW: 162.02 g/mol Boiling Pt: 221.2 °C (760 mm Hg) Flash Pt: 108.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD08704813 CAS Number: 56961-78-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...