4-Cyano-3-fluorophenylboronic acid 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
4-Cyano-3-fluorobenzeneboronic acid
A183122-1G
A183122-25G
A183122-5G
A183122-100G
A183122-10G
A183122-250MG
103815-610EA
6
USD
103815-610
103846-172
77083-762
77083-764
77083-766
77111-480
77111-482
77111-484
4-Cyano-3-fluorophenylboronic acid 97%
4-Cyano-3-fluorophenylboronic acid
Caution: for Research Use Only
Formula:
C₇H₅BFNO₂ MW: 164.93 g/mol Melting Pt: 362…366 °C Flash Pt: 168.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD03411549 CAS Number: 843663-18-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...