Fmoc-3-(1-naphthyl)-L-alanine 98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-1-Nal-OH

A182840-100G A182840-10G A182840-1G A182840-25G A182840-5G
103815-574EA 6 USD
103815-574 103846-134 76998-740 76998-742 77083-702 77083-704 77083-706
Fmoc-3-(1-naphthyl)-L-alanine 98%
Fmoc-3-(1-naphthyl)-L-alanine

Caution: for Research Use Only

Formula: C₂₈H₂₃NO₄
MW: 437.495 g/mol
Boiling Pt: 690.7 °C (760 mm Hg)
Melting Pt: 171…183 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00151914
CAS Number: 96402-49-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR