2,4,6-Tri-tert-butylaniline 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
2,4,6-Tris(tert-butyl)aniline
A178570-5G
A178570-100G
A178570-100MG
A178570-10G
A178570-250MG
A178570-25G
103807-354EA
14
USD
103807-354
103845-690
77083-122
77248-200
77248-202
77248-204
77248-206
77301-564
2,4,6-Tri-tert-butylaniline 98%
2,4,6-Tri-tert-butylaniline
Caution: for Research Use Only
Formula:
C₁₈H₃₁N Boiling Pt: 302 °C (760 mm Hg) Melting Pt: 145…147 °C Flash Pt: 123.7 °C Storage Temperature: Ambient |
MDL Number:
MFCD00011645 CAS Number: 961-38-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...