1,3-Dibromoadamantane 97%

Supplier: Ambeed
Warning

A174152-1G A174152-100G A174152-10G A174152-25G A174152-5G
103806-922EA 6 USD
103806-922 103845-220 77082-508 77191-512 77191-514 77301-320 77301-322
1,3-Dibromoadamantane 97%
1,3-Dibromoadamantane

Caution: for Research Use Only

Formula: C₁₀H₁₄Br₂
Boiling Pt: 287.6 °C (760 mm Hg)
Melting Pt: 112…113 °C
Flash Pt: 145.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00077218
CAS Number: 876-53-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR