D-Cycloserine 98%

Supplier: Ambeed
Warning

A170761-25G A170761-5G A170761-100MG A170761-10G A170761-1G A170761-100G
103844-814EA 9 USD
103844-814 77081-928 77081-930 77286-836 77286-838 77286-840 77567-508
D-Cycloserine 98%
D-Cycloserine

Caution: For research use only.

Formula: C₃H₆N₂O₂
Melting Pt: 147 °C
Storage Temperature: Freezer
MDL Number: MFCD00005353
CAS Number: 68-41-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR