5-Fluoroindole 98% (by GC)
Supplier: Ambeed
|
Warning
|
Synonyms:
5-Fluoro-1H-indole
A170102-1G
A170102-25G
A170102-5G
A170102-100G
A170102-10G
A170102-500G
103806-450EA
6
USD
103806-450
103844-716
77081-814
77081-816
77081-818
77273-668
77273-670
77273-672
5-Fluoroindole 98% (by GC)
5-Fluoroindole
Caution: for Research Use Only
Formula:
C₈H₆FN Boiling Pt: 258 °C (760 mm Hg) Melting Pt: 45…48 °C Flash Pt: 110 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00005671 CAS Number: 399-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...