Anti-PDX1 Rabbit Polyclonal Antibody

Supplier: ANTIBODIES.COM LLC
A97975-100
77060-454EA 484.2 USD
77060-454
Anti-PDX1 Rabbit Polyclonal Antibody
Antibodies
Rabbit polyclonal antibody to PDX1 for WB and ELISA with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:40000

Type: Primary
Antigen: PDX1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR