Anti-B2M Mouse Monoclonal Antibody [clone: KT134]

Supplier: ANTIBODIES.COM LLC
A185-100 A185-200
77041-674EA 504.6 USD
77041-674 77041-676
Anti-B2M Mouse Monoclonal Antibody [clone: KT134]
Antibodies
Mouse monoclonal (KT134) antibody to B2M for ELISA, WB and sELISA with samples derived from Human.

  • Validated Applications: ELISA, WB, sELISA
  • Recommended Dilutions: ELISA: 1/1000, WB: 1/500 to 1/10000, sELISA: 3 µg/ml

Type: Primary
Antigen: B2M
Clonality: Monoclonal
Clone: KT134
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR