Digital Cel-Gro Tissue Culture Rotator, Thermo Scientific

Supplier: Thermo Fisher Scientific
88882015
76486-002EA 2230.73 USD
76486-002 77041-358
Digital Cel-Gro Tissue Culture Rotator, Thermo Scientific
Shakers and Mixers Rotating/Rocking Shakers
The Digital Cel-Gro Tissue Culture Rotator has a brushless no-maintenance motor that provides a gentle rotating motion for optimum results.

  • Digital display of speed and time
  • Speed range: 5 to 70 rpm
  • Speed accuracy: ±1 rpm
  • Adjustable rocking angle: 95° to 180°
  • Loading capacity: 5 kg
  • Timer range: 0 to 9999 minutes
  • Optional multiple size drums offering different capacity of tube rotation

The Thermo Scientific™ Digital Cel-Gro Tissue Culture Rotator precisely mixes test tube contents. Achieve optimum culture results for tissue and virus growth, hormone production, and sample testing of viruses and cytotoxicity assays.

Certifications: cCSAus.

Ordering information: Order minimum 1 drum together with the rotator. 2 years of warranty.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR