Basic Vortex Mixer, Thermo Scientific

Supplier: Thermo Fisher Scientific
88882011
76485-986EA 493.2 USD
76485-986 77041-354
Basic Vortex Mixer, Thermo Scientific
Shakers and Mixers Vortex Mixers
Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.

  • Digital speed display
  • Continuous and touch modes
  • Speed range: 200 to 3000 rpm
  • Speed accuracy: ±5% rpm
  • Convenient speed adjustments by knob control
  • Stable for long-term operations
  • Multiple accessories for different vessels

Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.

Motor type: DC motor; Orbit: 4 mm; Material: ABS housing

Certifications: cCSAus.

Ordering information: 2 years of warranty.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR