ABT-737 98%

Supplier: Ambeed
Warning

A821402-100MG A821402-1MG A821402-5MG A821402-10MG
77036-542EA 429 USD
77036-542 77036-544 77036-546 77670-164
ABT-737 98%
ABT-737

Caution: for Research Use Only

Formula: C₄₂H₄₅ClN₆O₅S₂
MDL Number: MFCD12756212
CAS Number: 852808-04-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR