Fmoc-3-(trifluoromethyl)-L-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: Fmoc-Phe(3-CF₃)-OH, Fmoc-Phe(3-CF3)-OH, (S)-2-(9H-Fluoren-9-ylmethoxycarbonylamino)-3-(3-trifluoromethylphenyl)propionic acid, Fmoc-L-Phe-(3-trifluoromethyl)-OH, Fmoc-L-Phe-(3-CF3)-OH, 3-(Trifluoromethyl)-L-phenylalanine, N-FMOC protected

A749201-5G
77036-122EA 194.15 USD
77036-122
Fmoc-3-(trifluoromethyl)-L-phenylalanine 95%
Fmoc-3-(trifluoromethyl)-L-phenylalanine

Caution: for Research Use Only

Formula: C₂₅H₂₀F₃NO₄
MW: 455.44 g/mol
MDL Number: MFCD00672555
CAS Number: 205526-27-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR