2,2'-Bipyridyl 97%
Supplier: Ambeed
|
Danger
|
Synonyms:
α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl
A510112-25G
A510112-100G
A510112-10G
A510112-500G
A510112-5G
103839-482EA
5
USD
103839-482
76843-470
77034-580
77034-582
77034-584
77034-586
77307-656
2,2'-Bipyridyl 97%
2,2'-Bipyridyl
Caution: for Research Use Only
Formula:
C₁₀H₈N₂ Boiling Pt: 272.5 °C (760 mm Hg) Melting Pt: 70…73 °C Flash Pt: 121 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006212 CAS Number: 366-18-7 UN: 2811 ADR: 6.1,Ⅲ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...