2,2'-Bipyridyl 97%

Supplier: Ambeed
Danger

Synonyms: α,α′-Dipyridyl, 2,2'-Bipyridine, 2,2'-Dipyridyl

A510112-25G A510112-100G A510112-10G A510112-500G A510112-5G
103839-482EA 5 USD
103839-482 76843-470 77034-580 77034-582 77034-584 77034-586 77307-656
2,2'-Bipyridyl 97%
2,2'-Bipyridyl

Caution: for Research Use Only

Formula: C₁₀H₈N₂
Boiling Pt: 272.5 °C (760 mm Hg)
Melting Pt: 70…73 °C
Flash Pt: 121 °C
Storage Temperature: Ambient
MDL Number: MFCD00006212
CAS Number: 366-18-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR