PLX4720 99+%
Supplier: Ambeed
|
Warning
|
Synonyms:
N-(3-(5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulfonamide, PLX-4720, N-(3-(5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulphonamide
A332013-10MG
A332013-1MG
A332013-25MG
A332013-50MG
A332013-5MG
A332013-250MG
A332013-100MG
77032-884EA
15
USD
77032-884
77032-886
77032-888
77032-890
77032-892
77668-504
77872-478
PLX4720 99+%
PLX4720
Caution: for Research Use Only
Formula:
C₁₇H₁₄ClF₂N₃O₃S |
MDL Number:
MFCD14635203 CAS Number: 918505-84-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...