PLX4720 99+%

Supplier: Ambeed
Warning

Synonyms: N-(3-(5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulfonamide, PLX-4720, N-(3-(5-Chloro-1H-pyrrolo[2,3-b]pyridine-3-carbonyl)-2,4-difluorophenyl)propane-1-sulphonamide

A332013-10MG A332013-1MG A332013-25MG A332013-50MG A332013-5MG A332013-250MG A332013-100MG
77032-884EA 15 USD
77032-884 77032-886 77032-888 77032-890 77032-892 77668-504 77872-478
PLX4720 99+%
PLX4720

Caution: for Research Use Only

Formula: C₁₇H₁₄ClF₂N₃O₃S
MDL Number: MFCD14635203
CAS Number: 918505-84-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR