Ruxolitinib 98%

Supplier: Ambeed
Warning

A272323-100MG A272323-10MG A272323-1G A272323-1MG A272323-250MG A272323-5MG A272323-5G
77031-962EA 56 USD
77031-962 77031-964 77031-966 77031-968 77031-970 77031-972 77872-321
Ruxolitinib 98%
Ruxolitinib

Caution: for Research Use Only

Formula: C₁₇H₁₈N₆
MDL Number: MFCD12031592
CAS Number: 941678-49-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR