Ruxolitinib 98%
Supplier: Ambeed
|
Warning
|
A272323-100MG
A272323-10MG
A272323-1G
A272323-1MG
A272323-250MG
A272323-5MG
A272323-5G
77031-962EA
56
USD
77031-962
77031-964
77031-966
77031-968
77031-970
77031-972
77872-321
Ruxolitinib 98%
Ruxolitinib
Caution: for Research Use Only
Formula:
C₁₇H₁₈N₆ |
MDL Number:
MFCD12031592 CAS Number: 941678-49-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...