Alizarin complexone 95%

Supplier: Ambeed
Warning

A235856-1G A235856-25G A235856-5G
77031-190EA 10 USD
77031-190 77031-192 77031-194 77031-196
Alizarin complexone 95%
Alizarin complexone

Caution: for Research Use Only

Formula: C₁₉H₁₅NO₈
MDL Number: MFCD00001202
CAS Number: 3952-78-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR