Glycitein 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
7-Hydroxy-3-(4-hydroxyphenyl)-6-methoxy-4H-chromen-4-one, 7,4'-Dihydroxy-6-methoxyisoflavone
A231798-1MG
A231798-5MG
77031-068EA
14
USD
77031-068
77031-070
Glycitein 98%
Glycitein
Caution: for Research Use Only
Formula:
C₁₆H₁₂O₅ MW: 284.27 g/mol |
MDL Number:
MFCD00016679 CAS Number: 40957-83-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...