Fmoc-Glu-OtBu 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(S)-4-((((9H-Fluoren-9-yl)methoxy)carbonyl)amino)-5-(tert-butoxy)-5-oxopentanoic acid, N-Fmoc-L-glutamic acid 1-tert-butyl ester, Fmoc-Glu-OtBu
A175101-100G
A175101-10G
A175101-500G
A175101-25G
A175101-5G
A175101-1G
A175101-1000G
103807-000EA
6
USD
103807-000
103845-304
77029-226
77029-228
77029-230
77082-626
77082-628
77301-364
77871-875
Fmoc-Glu-OtBu 98%
Fmoc-Glu-OtBu
Caution: for Research Use Only
Formula:
C₂₄H₂₇NO₆ MW: 425.47 g/mol Boiling Pt: 638.1 °C (760 mm Hg) Storage Temperature: Ambient |
MDL Number:
MFCD00065648 CAS Number: 84793-07-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...