N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98% (by HPLC) (max.6% H₂O)
Supplier: Ambeed
|
Warning
|
A171345-100G
A171345-10G
A171345-500G
A171345-5G
A171345-25G
77029-034EA
35
USD
77029-034
77029-036
77029-038
77029-040
77301-124
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate 98% (by HPLC) (max.6% H₂O)
N-[(9H-Fluoren-9-ylmethoxy)carbonyl]-D-alanine hydrate
Caution: for Research Use Only
Formula:
C₁₈H₁₇NO₄ |
MDL Number:
MFCD00062960 CAS Number: 79990-15-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...