Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 98%

Supplier: Ambeed
Warning

A168222-1000G A168222-100G A168222-500G A168222-25G
77028-908EA 59 USD
77028-908 77028-910 77028-912 77300-924
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol) 98%
Tris(hydroxymethyl)aminomethane (TRIS, Trometamol)

Caution: for Research Use Only

Formula: NH₂C(CH₂OH)₃
MDL Number: MFCD00004679
CAS Number: 77-86-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR