Cyclohexyldiphenylphosphine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Diphenylcyclohexylphosphine
A163530-100G
A163530-10G
A163530-25G
A163530-5G
103805-736EA
5
USD
103805-736
103843-996
77028-702
77028-704
77320-094
77320-096
Cyclohexyldiphenylphosphine 98%
Cyclohexyldiphenylphosphine
Caution: for Research Use Only
Formula:
C₁₈H₂₁P MW: 268.33 g/mol Boiling Pt: 374 °C (760 mm Hg) Melting Pt: 58…62 °C Flash Pt: 189.4 °C Storage Temperature: Ambient |
MDL Number:
MFCD00046360 CAS Number: 6372-42-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...