Cyclohexyldiphenylphosphine 98%

Supplier: Ambeed
Warning

Synonyms: Diphenylcyclohexylphosphine

A163530-100G A163530-10G A163530-25G A163530-5G
103805-736EA 5 USD
103805-736 103843-996 77028-702 77028-704 77320-094 77320-096
Cyclohexyldiphenylphosphine 98%
Cyclohexyldiphenylphosphine

Caution: for Research Use Only

Formula: C₁₈H₂₁P
MW: 268.33 g/mol
Boiling Pt: 374 °C (760 mm Hg)
Melting Pt: 58…62 °C
Flash Pt: 189.4 °C
Storage Temperature: Ambient
MDL Number: MFCD00046360
CAS Number: 6372-42-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR