Linifanib 98+%

Supplier: Ambeed
Warning

A147574-1MG A147574-50MG A147574-5MG A147574-250MG
77027-968EA 19 USD
77027-968 77027-970 77027-972 77871-721
Linifanib 98+%
Linifanib

Caution: for Research Use Only

Formula: C₂₁H₁₈FN₅O
MDL Number: MFCD11840918
CAS Number: 796967-16-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR