2-Bromoisonicotinonitrile 95%
Supplier: Ambeed
|
Warning
|
A137674-100G
A137674-10G
A137674-1G
A137674-25G
A137674-5G
103802-482EA
5
USD
103802-482
103833-470
77027-358
77027-360
77315-456
77315-458
77315-460
2-Bromoisonicotinonitrile 95%
2-Bromoisonicotinonitrile
Caution: for Research Use Only
Formula:
C₆H₃BrN₂ Boiling Pt: 243 °C (760 mm Hg) Melting Pt: 78…80 °C Flash Pt: 101 °C Storage Temperature: Ambient |
MDL Number:
MFCD07367879 CAS Number: 10386-27-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...