Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]

Supplier: AAT Bioquest
10041000
77022-750EA 164.21 USD
77022-750
Anti-CD4 Mouse Monoclonal Antibody [clone: RPA-T4]
Antibodies
RPA-T4 is an anti-human monoclonal antibody that targets the CD4 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Primary
Antigen: CD4
Clonality: Monoclonal
Clone: RPA-T4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR