Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet)

Supplier: AAT BIOQUEST INC
103400Z0 103400Z1
77025-014EA 244.24 USD
77025-014 77021-390
Anti-CD34 Mouse Monoclonal Antibody [clone: 4H11] (mFluor™ Violet)
Antibodies
4H11 is an anti-human monoclonal antibody that recognizes the CD34 antigen.

  • Purified through affinity chromatography

Commonly used in flow cytometry applications as a phenotypic marker for differentiation of cell types.

Caution: For research use only.

Type: Secondary
Antigen: CD34
Clonality: Monoclonal
Clone: 4H11
Conjugation: mFluor™ Violet
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR