PTS Panels® Total Cholesterol Test Strips
Supplier: PTS Diagnostics
1711
77020-340EA 138.85 USD
77020-340
PTS Panels® Total Cholesterol Test Strips
Clinical Diagnostic Supplies Clinical Reagents and Kits
PTS Panels Cholesterol test strips provide lab-quality total cholesterol results on-site in about 45 seconds from a single fingerstick.
  • Measures total cholesterol
  • 25 test strips per vial – Professional
  • Refrigeration not required
PTS Panels Cholesterol test strips are for professional use with CardioChek PA and CardioChek Plus analyzers to measure total cholesterol in venous whole blood or capillary fingerstick blood.
Cholesterol measurements are used in the diagnosis and treatments of disorders involving excess cholesterol in the blood and lipid and lipoprotein metabolism disorders. Lipoprotein measurements are used in the diagnosis and treatment of lipid disorders (such as diabetes mellitus), atherosclerosis, and various liver and renal diseases.
This test system will detect cholesterol levels from 100 to 400 mg/dl (2.59 to 10.36 mmol/L) and will display a number value for results in this range.