CardioChek PA Blood Sample Analyzer
Supplier: PTS Diagnostics
1708
77020-338EA 987.89 USD
77020-338
CardioChek PA Blood Sample Analyzer
Clinical Diagnostic Supplies Clinical Analysers
The CardioChek PA analyzer achieves accuracy similar to that of a clinical laboratory, but only requires a small (40 μl) blood sample from a fingerstick.
  • Compact, battery-operated
  • Meets NCEP guidelines for accuracy and precision
  • Room temperature test strip storage
  • CLIA Waved
Full lipid panel results in as little as 90 seconds. Single blood chemistry in as little as 45 seconds.
Certifications: CLIA-waived.