Zymo-Spin II-PX Column
Supplier: Zymo Research
C1086-50
77020-324PK 74 USD
77020-324
Zymo-Spin II-PX Column
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA.
  • Allows for purification of up to 100 µg of plasmid DNA in ≥25 µl eluate when used in combination with ZymoPURE™ plasmid prep buffers
Available as a refill for the ZymoPURE™ plasmid miniprep kit. The versatile Zymo-Spin II-PX column can be used in microcentrifuges or on vacuum manifolds for the purification of plasmid DNA. The Zymo-Spin II-PX features durable polypropylene construction and a unique silica-based matrix. Capacity is 800 µl.