DNA/RNA Shield Fecal Collection Tube (with Beads)
Supplier: Zymo Research
R1137
77020-304PK 95.09 USD
77020-304
DNA/RNA Shield Fecal Collection Tube (with Beads)
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
DNA/RNA shield fecal collection tubes are designed for the collection and preservation of nucleic acids from stool specimens.
  • Pre-filled with DNA/RNA shield to inactivate pathogens and stabilize microbiome profiles (DNA and RNA) at ambient temperature
DNA/RNA Shield Fecal Collection tubes are designed for the collection and preservation of nucleic acids from stool specimens. These fecal collection tubes take a microbial snapshot of a sample while inactivating viruses making samples safe and ready for transport. Samples stored in these fecal collection tubes are stable at ambient temperature, and can be frozen for longer-term storage. Simply collect a spoonful of the specimen into the stool collection tubes prefilled with preservative and shake vigorously to ensure proper stabilization. R1137 is pre-filled with 5 glass beads to help facilitate homogenization.