Zymo-Spin VI-PX Column
Supplier: Zymo Research
C1080-5
77020-294PK 146.71 USD
77020-294
Zymo-Spin VI-PX Column
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The versatile Zymo-Spin VI-PX can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA.
  • Exclusive to this column is a Luer-Lock bottom assembly and conical tip
The Zymo-Spin VI-PX features durable polypropylene construction and a unique silica-based matrix that allows for purification of up to 25 mg of plasmid DNA in ≥3 ml eluate when used in combination with ZymoPURE™ Plasmid Prep buffers. Capacity is 15 ml and can be increased to 615 ml when used in conjunction with the 600 ml reservoir.