Zymo-Spin VP-X Column
Supplier: Zymo Research
C1082-5
77020-278PK 97.02 USD
77020-278
Zymo-Spin VP-X Column
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The versatile Zymo-Spin V-PX with 15 and 50 ml reservoirs can be used in centrifuges or on vacuum manifolds for the purification of plasmid DNA.
  • Allows for purification of up to 3 mg of plasmid DNA in ≥300 µl eluate when used in combination with ZymoPURE™ plasmid prep buffers
Available as a refill for the ZymoPURE™ II plasmid maxiprep kit. The Zymo-Spin V-PX features durable polypropylene construction and a unique silica-based matrix. Capacity with reservoir assembly is 65 ml.