ZymoPURE™ Filter Plate
Supplier: Zymo Research
C2022
77020-268EA 165.14 USD
77020-268 77020-288
ZymoPURE™ Filter Plate
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
The ZymoPURE™ Filter Plate can be used with centrifuges and vacuum manifolds for the rapid clarification of bacterial lysates.
  • The capacity of the well is 1.0 ml
Available as a refill for the ZymoPURE™ 96 Plasmid Miniprep Kit. The ZymoPURE™ Filter Plate can be used with centrifuges and vacuum manifolds for the rapid clarification of bacterial lysates when used in conjunction with our ZymoPURE™ Plasmid Prep buffers.