ZymoScript RT PreMix Kit
Supplier: Zymo Research
R3012
77020-264EA 421.48 USD
77020-264
ZymoScript RT PreMix Kit
Nucleic Acid Reagents Nucleic Acid Purification Kits and Reagents
ZymoScript RT PreMix is an optimized reverse transcription kit containing all the necessary components for quick and efficient cDNA synthesis.
  • Efficient and quick cDNA synthesis (in as little as 10 minutes)
ZymoScript RT PreMix is an optimized reverse transcription kit containing all the necessary components for quick and efficient cDNA synthesis. This reverse transcription mix includes random hexamer and oligo dT primers, dNTPs, RNase inhibitors, reverse transcriptase, and an optimized reaction buffer which includes a visual tracking dye for accurate reaction set-up. This is the kit of choice for analysis of RNA via 2-step RT-qPCR.