2-Bromopyridine 99%

Supplier: Ambeed
Warning

A739104-25G A739104-1000G A739104-100G A739104-500G
76845-050EA 14 USD
76845-050 77018-942 77018-944 77018-946
2-Bromopyridine 99%
2-Bromopyridine

Caution: for Research Use Only

Formula: C₅H₄BrN
MDL Number: MFCD00006219
CAS Number: 109-04-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR