6-Thioguanine 98%

Supplier: Ambeed
Danger

A393548-100MG A393548-1G A393548-250MG A393548-25MG A393548-5G
77016-220EA 7 USD
77016-220 77016-222 77016-224 77016-226 77572-196
6-Thioguanine 98%
6-Thioguanine

Caution: for Research Use Only

Formula: C₅H₅N₅S
MDL Number: MFCD00233553
CAS Number: 154-42-7
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR