2-Amino-6-chloro-3-nitropyridine 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine
A304437-100G
A304437-10G
A304437-250MG
A304437-25G
A304437-500G
77015-000EA
38
USD
77015-000
77015-002
77015-004
77015-006
77015-008
77015-010
2-Amino-6-chloro-3-nitropyridine 98%
2-Amino-6-chloro-3-nitropyridine
Caution: for Research Use Only
Formula:
C₅H₄ClN₃O₂ MW: 173.56 |
MDL Number:
MFCD00044350 CAS Number: 27048-04-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...