2-Amino-6-chloro-3-nitropyridine 98%

Supplier: Ambeed
Warning

Synonyms: 6-Chloro-3-nitro-2-pyridylamine, 6-Chloro-3-nitropyridin-2-amine

A304437-100G A304437-10G A304437-250MG A304437-25G A304437-500G
77015-000EA 38 USD
77015-000 77015-002 77015-004 77015-006 77015-008 77015-010
2-Amino-6-chloro-3-nitropyridine 98%
2-Amino-6-chloro-3-nitropyridine

Caution: for Research Use Only

Formula: C₅H₄ClN₃O₂
MW: 173.56
MDL Number: MFCD00044350
CAS Number: 27048-04-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR