Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A181309-1G A181309-250MG A181309-5G A181309-100MG
77012-006EA 227 USD
77012-006 77012-008 77567-662 77683-455
Sulfo-SMCC (3-Sulfo-N-succinimidyl-4-(N-maleimidomethyl)cyclohexane-1-carboxylate sodium salt) 98%, Ambeed.Inc

Caution: For research use only.

Formula: C₁₆H₁₈N₂O₉S
MDL Number: MFCD00054978
CAS Number: 92921-24-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR